Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3-[2-[2-(2-mercaptoethoxy)ethoxy]ethoxy]- | 1347750-82-6 | ≥95.0% | 238.30 | 1 G
SDP

Catalog No. 5000349850
Encompass_Preferred

Thiol-PEG3-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker; one ligand binds to an E3 ubiquitin ligase, and the other to the target protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Formula: C9H18O5S
  • Purity: ≥95.0%
  • Molecular weight: 238.30
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-49850 Supplier Medchemexpress LLC Supplier No. HY1400181G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-PEG3-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs consist of two distinct ligands joined by a linker; one ligand binds to an E3 ubiquitin ligase, and the other to the target protein. PROTACs function by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Formula: C9H18O5S
  • Purity: ≥95.0%
  • Molecular weight: 238.30
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.