Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3-[2-[2-[(4-formylbenzoyl)amino]ethoxy]ethoxy]-, 2,5-dioxo-1-pyrrolidinyl ester | 1807521-07-8 | 98.5% | 406.39 | C19H22N2O8 | 50 MG
SDP

Catalog No. 5000202474
Encompass_Preferred

Ald-Ph-amido-PEG2-C2-NHS ester is a non-cleavable 2-unit PEG linker used for antibody-drug conjugation (ADC). It features an N-hydroxysuccinimide (NHS) ester for amine-reactive coupling and a 4-formylbenzoyl aldehyde handle for further derivatization. Supplied as a colorless to light yellow liquid, the material is validated by 1H NMR and LCMS.

  • Non-cleavable 2-unit polyethylene glycol (PEG) linker for ADC chemistry.
  • N-hydroxysuccinimide (NHS) ester for amine-reactive conjugation.
  • 4-formylbenzoyl aldehyde for further derivatization.
  • Molecular formula C19H22N2O8; molecular weight 406.39.
  • CAS number 1807521-07-8.
  • Purity (LCMS) 98.5%.
  • Appearance colorless to light yellow liquid.
  • Storage: pure form -20°C up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Pack size typically supplied as 50 MG.

Catalog No. 50-002-02474 Supplier Medchemexpress LLC Supplier No. HY13010450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ald-Ph-amido-PEG2-C2-NHS ester is a non-cleavable 2-unit PEG linker used for antibody-drug conjugation (ADC). It features an N-hydroxysuccinimide (NHS) ester for amine-reactive coupling and a 4-formylbenzoyl aldehyde handle for further derivatization. Supplied as a colorless to light yellow liquid, the material is validated by 1H NMR and LCMS.

  • Non-cleavable 2-unit polyethylene glycol (PEG) linker for ADC chemistry.
  • N-hydroxysuccinimide (NHS) ester for amine-reactive conjugation.
  • 4-formylbenzoyl aldehyde for further derivatization.
  • Molecular formula C19H22N2O8; molecular weight 406.39.
  • CAS number 1807521-07-8.
  • Purity (LCMS) 98.5%.
  • Appearance colorless to light yellow liquid.
  • Storage: pure form -20°C up to 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
  • Pack size typically supplied as 50 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.