Learn More
Medchemexpress LLC Propanoic acid, 3-[2-[2-[(4-formylbenzoyl)amino]ethoxy]ethoxy]-, 2,5-dioxo-1-pyrrolidinyl ester | 1807521-07-8 | 98.5% | 406.39 g·mol⁻1 | C19H22N2O8 | 100 MG
SDP
Catalog No. 5000204110
Encompass_Preferred
Ald-Ph-amido-PEG2-C2-NHS ester is a noncleavable two-unit polyethylene glycol (PEG) linker designed for antibody-agent conjugation (ADC) research. It features an NHS-activated ester for amine coupling and an aldehyde-phenyl functional handle for orthogonal conjugation chemistry.
  • Provides a noncleavable 2-unit PEG spacer for conjugation
  • NHS ester activated for efficient coupling to primary amines
  • Aldehyde-phenyl functional group for orthogonal conjugation chemistry
  • High purity suitable for research applications
  • Available in multiple pack sizes and formats, including 100 MG
Catalog No. 50-002-04110 Supplier Medchemexpress LLC Supplier No. HY130104100MG
Ald-Ph-amido-PEG2-C2-NHS ester is a noncleavable two-unit polyethylene glycol (PEG) linker designed for antibody-agent conjugation (ADC) research. It features an NHS-activated ester for amine coupling and an aldehyde-phenyl functional handle for orthogonal conjugation chemistry.
  • Provides a noncleavable 2-unit PEG spacer for conjugation
  • NHS ester activated for efficient coupling to primary amines
  • Aldehyde-phenyl functional group for orthogonal conjugation chemistry
  • High purity suitable for research applications
  • Available in multiple pack sizes and formats, including 100 MG