Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-azidoethoxy)ethoxy]-, 1,1-dimethylethyl ester. | 1271728-79-0 | MFCD22384661 | 95.0% | C11H21N3O4 | 500 MG
SDP
Catalog No. 5000355693
Encompass_Preferred
Azido-PEG2-C2-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This reagent facilitates click chemistry reactions, featuring an azide group that readily participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups. It also engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules possessing DBCO or BCN groups, offering versatility in chemical modifications.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Click chemistry reagent with an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
Catalog No. 50-003-55693 Supplier Medchemexpress LLC Supplier No. HY140776500MG
Azido-PEG2-C2-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This reagent facilitates click chemistry reactions, featuring an azide group that readily participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups. It also engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules possessing DBCO or BCN groups, offering versatility in chemical modifications.
  • PEG-based PROTAC linker.
  • Used in the synthesis of PROTACs.
  • Click chemistry reagent with an azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.