Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-bromoethoxy)ethoxy]-, 1,1-dimethylethyl ester | 1381861-91-1 | 97.0% | C11H21BrO4 | 1 G
SDP
Catalog No. 5000356304
Encompass_Preferred
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two different ligands connected by a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Colorless to light yellow liquid appearance
  • Purity of 97.0%
Catalog No. 50-003-56304 Supplier Medchemexpress LLC Supplier No. HY1413651G
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins, featuring two different ligands connected by a linker.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Colorless to light yellow liquid appearance
  • Purity of 97.0%