Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-bromoethoxy)ethoxy]-, 1,1-dimethylethyl ester | 1381861-91-1 | 97.0% | C11H21BrO4 | 25 G
SDP
Catalog No. 5000352604
Encompass_Preferred
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This compound is suitable for research applications requiring targeted protein degradation.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligands for an E3 ubiquitin ligase and a target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52604 Supplier Medchemexpress LLC Supplier No. 25GHY141365
Bromo-PEG2-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This compound is suitable for research applications requiring targeted protein degradation.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Ligands for an E3 ubiquitin ligase and a target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins