Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-propyn-1-yloxy)ethoxy]-, 1,1-dimethylethyl ester | 1807503-80-5 | 95.0% | 1 G
SDP
Catalog No. 5000341071
Encompass_Preferred
Propargyl-PEG2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Functions as a PEG-based PROTAC linker
  • Used for synthesizing PROTACs
  • Acts as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules
Catalog No. 50-003-41071 Supplier Medchemexpress LLC Supplier No. HY1305971G
Propargyl-PEG2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Alkyne group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Functions as a PEG-based PROTAC linker
  • Used for synthesizing PROTACs
  • Acts as a click chemistry reagent
  • Contains an Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules