Learn More
Medchemexpress LLC Propanoic acid, 3-[2-(2-propyn-1-yloxy)ethoxy]-, 1,1-dimethylethyl ester | 1807503-80-5 | 95.0% | 250 MG
SDP
Catalog No. 5000341762
Encompass_Preferred
This compound is a polyethylene glycol (PEG)-based PROTAC linker designed for use in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It acts as a click chemistry reagent due to its alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Application: Can be employed in the synthesis of PROTACs.
  • Characteristics: It contains an alkyne group and can participate in copper-catalyzed azide-alkyne cycloaddition.
Catalog No. 50-003-41762 Supplier Medchemexpress LLC Supplier No. 250MGHY130597
This compound is a polyethylene glycol (PEG)-based PROTAC linker designed for use in the synthesis of Proteolysis Targeting Chimeras (PROTACs). It acts as a click chemistry reagent due to its alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition with azide-containing molecules.
  • Application: Can be employed in the synthesis of PROTACs.
  • Characteristics: It contains an alkyne group and can participate in copper-catalyzed azide-alkyne cycloaddition.