Learn More
Medchemexpress LLC Propanoic acid, 3-(2-azidoethoxy)-, 2,5-dioxo-1-pyrrolidinyl ester | 1807530-06-8 | 99.8% | C9H12N4O5 | 100 MG
SDP
Catalog No. 5000353544
Encompass_Preferred
Azido-PEG1-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Acts as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups
Catalog No. 50-003-53544 Supplier Medchemexpress LLC Supplier No. HY140756100MG
Azido-PEG1-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Functions as a PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Acts as a click chemistry reagent
  • Contains an azide group for chemical reactions
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups