Learn More
Medchemexpress LLC Propanoic acid, 3-(2-bromoethoxy)- | 1393330-33-0 | 98.0% | 100 MG
SDP
Catalog No. 5000338363
Encompass_Preferred
Bromo-PEG1-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein, leveraging the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • High purity of 98.0%
  • Appears as a liquid, ranging from light yellow to yellow
  • Requires storage at -20°C, protected from light
Catalog No. 50-003-38363 Supplier Medchemexpress LLC Supplier No. HY130425100MG
Bromo-PEG1-acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein, leveraging the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • Functions as a PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • High purity of 98.0%
  • Appears as a liquid, ranging from light yellow to yellow
  • Requires storage at -20°C, protected from light