Learn More
Medchemexpress LLC Propanoic acid, 3-(2-mercaptoethoxy)- | 1260092-48-5 | 95.0% | 150.20 g/mol | C5H10O3S | 50 MG
SDP
Catalog No. 5000283190
Encompass_Preferred
Thiol-PEG1-acid (SH-PEG1-COOH) is a short polyethylene glycol derivative bearing a reactive thiol (-SH) and a terminal carboxylic acid (-COOH). It is supplied as a colorless to light yellow liquid and is intended for use as a linker in bioconjugation and PEGylation reactions. Store pure material frozen and protect solutions from long-term degradation.
Catalog No. 50-002-83190 Supplier Medchemexpress LLC Supplier No. HYW19087750MG
Add to Cart
Add to Cart
Thiol-PEG1-acid (SH-PEG1-COOH) is a short polyethylene glycol derivative bearing a reactive thiol (-SH) and a terminal carboxylic acid (-COOH). It is supplied as a colorless to light yellow liquid and is intended for use as a linker in bioconjugation and PEGylation reactions. Store pure material frozen and protect solutions from long-term degradation.