Learn More
Medchemexpress LLC Propanoic acid, 3,3'-[(2-amino-1,3-propanediyl)bis(oxy)]bis-, 1,1'-bis(1,1-dimethylethyl) ester | 2171072-53-8 | 99.7% | C17H33NO6 | 250 MG
SDP
Catalog No. 5000357230
Encompass_Preferred
NH-bis(C1-PEG1-Boc) is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. This compound is a key component in creating PROTACs, which are molecules designed to selectively degrade target proteins within a cell.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-57230 Supplier Medchemexpress LLC Supplier No. 250MGHY140252
NH-bis(C1-PEG1-Boc) is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. This compound is a key component in creating PROTACs, which are molecules designed to selectively degrade target proteins within a cell.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins