Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, 3,3'-[(2-amino-1,3-propanediyl)bis(oxy)]bis-, 1,1'-bis(1,1-dimethylethyl) ester | 2171072-53-8 | 99.7% | C17H33NO6 | 50 MG
SDP

Catalog No. 5000353007
Encompass_Preferred

NH-bis(C1-PEG1-Boc) from MedChemExpress is a chemical compound with the molecular formula C17H33NO6 and a molecular weight of 347.45. It is stored at 4°C, sealed, and protected from moisture and light. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month under similar conditions.

  • Appearance: Colorless to light yellow liquid
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure
  • Purity of 99.72% by HPLC

Catalog No. 50-003-53007 Supplier Medchemexpress LLC Supplier No. HY14025250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NH-bis(C1-PEG1-Boc) from MedChemExpress is a chemical compound with the molecular formula C17H33NO6 and a molecular weight of 347.45. It is stored at 4°C, sealed, and protected from moisture and light. In solvent, it can be stored at -80°C for 6 months or -20°C for 1 month under similar conditions.

  • Appearance: Colorless to light yellow liquid
  • 1H NMR spectrum consistent with structure
  • LCMS consistent with structure
  • Purity of 99.72% by HPLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.