Learn More
Medchemexpress LLC Propanoic acid, 3-bromo-2-oxo- | 1113-59-3 | ≥98.0% | 166.96 | 10 G
SDP
Catalog No. 5000405005
Encompass_Preferred
3-Bromopyruvic acid, also known as bromopyruvic acid or 3-BrPA, is a pyruvate analogue that acts as a potent hexokinase (HK)-II inhibitor. It demonstrates high tumor selectivity, inhibiting cell growth and inducing apoptosis by disrupting glycolysis. This compound also promotes autophagy through stimulating ROS formation in breast cancer cells and exhibits antimicrobial properties.
  • Potent hexokinase (HK)-II inhibitor
  • High tumor selectivity
  • Inhibits cell growth
  • Induces apoptosis by interfering with glycolysis
  • Induces autophagy by stimulating ROS formation in breast cancer cells
  • Exhibits antimicrobial activities
Catalog No. 50-004-05005 Supplier Medchemexpress LLC Supplier No. HY1999210G
3-Bromopyruvic acid, also known as bromopyruvic acid or 3-BrPA, is a pyruvate analogue that acts as a potent hexokinase (HK)-II inhibitor. It demonstrates high tumor selectivity, inhibiting cell growth and inducing apoptosis by disrupting glycolysis. This compound also promotes autophagy through stimulating ROS formation in breast cancer cells and exhibits antimicrobial properties.
  • Potent hexokinase (HK)-II inhibitor
  • High tumor selectivity
  • Inhibits cell growth
  • Induces apoptosis by interfering with glycolysis
  • Induces autophagy by stimulating ROS formation in breast cancer cells
  • Exhibits antimicrobial activities