Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propanoic acid, PEG4-NHPI pentafluorophenyl ester | 2101206-46-4 | >97.0% | 577.45 | C25H24F5NO9 | 100 MG
SDP

Catalog No. 5000202617
Encompass_Preferred

NHPI-PEG4-C2-Pfp ester is a PEG4-based pentafluorophenyl-activated linker reagent used for antibody-drug conjugate synthesis and other bioconjugation applications.

  • Used as a linker reagent for antibody-drug conjugates and bioconjugation.
  • High purity (≥97.0% by NMR).
  • Molecular weight 577.45; chemical formula C25H24F5NO9.
  • Colorless to light yellow liquid appearance.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-02617 Supplier Medchemexpress LLC Supplier No. HY130090100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NHPI-PEG4-C2-Pfp ester is a PEG4-based pentafluorophenyl-activated linker reagent used for antibody-drug conjugate synthesis and other bioconjugation applications.

  • Used as a linker reagent for antibody-drug conjugates and bioconjugation.
  • High purity (≥97.0% by NMR).
  • Molecular weight 577.45; chemical formula C25H24F5NO9.
  • Colorless to light yellow liquid appearance.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.