Learn More
Medchemexpress LLC Propanoic-d5 acid, sodium salt | 202529-18-8 | 99.9% | 10 MG
SDP
Catalog No. 5000297129
Encompass_Preferred
Propanoic-d5 (sodium) is the deuterium-labeled sodium salt of propionic acid used as a stable isotope tracer in analytical and metabolic studies.
  • Deuterium-labeled compound suitable as an internal standard or tracer in mass spectrometry and metabolic research.
  • High chemical purity (99.91% by HPLC) for reliable analytical performance.
  • Deuterium incorporation (d5) provides clear isotopic distinction from unlabeled analogs.
  • Solid form, facilitating handling and long-term storage.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-97129 Supplier Medchemexpress LLC Supplier No. HYB1773AS410MG
Propanoic-d5 (sodium) is the deuterium-labeled sodium salt of propionic acid used as a stable isotope tracer in analytical and metabolic studies.
  • Deuterium-labeled compound suitable as an internal standard or tracer in mass spectrometry and metabolic research.
  • High chemical purity (99.91% by HPLC) for reliable analytical performance.
  • Deuterium incorporation (d5) provides clear isotopic distinction from unlabeled analogs.
  • Solid form, facilitating handling and long-term storage.
  • Recommended storage: powder at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).