Learn More
Apexbio Technology LLC Propantheline bromide 50-34-0 100mg
SDP
Catalog No. 5000088389
Encompass_Preferred
Propantheline bromide (CAS 50-34-0) is a small-molecule antagonist of muscarinic acetylcholine receptors (mAChRs) It inhibits cholinergic neurotransmission within smooth muscle tissues resulting in modulation of muscle contractility In biomedical research propantheline bromide is utilized to investigate physiological mechanisms related to smooth muscle disorders such as gastrointestinal or bladder spasms and hyperhidrosis This compound is broadly applied in studies of the autonomic nervous system and serves as a tool for pharmacological exploration of cholinergic signaling pathways
Catalog No. 50-000-88389 Supplier Apexbio Technology LLC Supplier No. C6594100
Propantheline bromide (CAS 50-34-0) is a small-molecule antagonist of muscarinic acetylcholine receptors (mAChRs) It inhibits cholinergic neurotransmission within smooth muscle tissues resulting in modulation of muscle contractility In biomedical research propantheline bromide is utilized to investigate physiological mechanisms related to smooth muscle disorders such as gastrointestinal or bladder spasms and hyperhidrosis This compound is broadly applied in studies of the autonomic nervous system and serves as a tool for pharmacological exploration of cholinergic signaling pathways