Learn More
TARGETMOL CHEMICALS INC Propantheline bromide 500MG
SDP
Catalog No. 502880350
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Propantheline bromide (Pro-Banthine) competitively antagonizes acetylcholine activity mediated by muscarinic receptors at neuroeffector sites on smooth muscle and exocrine gland cells. Propantheline Bromide is the bromide salt form of propantheline, a quaternary ammonium compound structurally related to belladonna alkaloids. An aspartic acid residue present in the N-terminal portion of the third transmembrane helix of the muscarinic receptor is believed to form an ionic bond with the tertiary or quaternary nitrogen of the antagonist. Antagonism leads to a reduction of exocrine glands secretions and to relax the bronchial muscle and reduce tone and motility of intestinal smooth muscle. Purity 95%
Catalog No. 50-288-0350 Supplier TARGETMOL CHEMICALS INC Supplier No. T1006500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Propantheline bromide (Pro-Banthine) competitively antagonizes acetylcholine activity mediated by muscarinic receptors at neuroeffector sites on smooth muscle and exocrine gland cells. Propantheline Bromide is the bromide salt form of propantheline, a quaternary ammonium compound structurally related to belladonna alkaloids. An aspartic acid residue present in the N-terminal portion of the third transmembrane helix of the muscarinic receptor is believed to form an ionic bond with the tertiary or quaternary nitrogen of the antagonist. Antagonism leads to a reduction of exocrine glands secretions and to relax the bronchial muscle and reduce tone and motility of intestinal smooth muscle. Purity 95%