Learn More
Medchemexpress LLC Propantheline-d3 bromide | 64717-35-7 | 99.0% | 1 MG
SDP
Catalog No. 5000411285
Encompass_Preferred
Propantheline-d3 (bromide) is a deuterium-labeled version of Propantheline bromide. Propantheline bromide is an antimuscarinic agent used to treat hyperhidrosis, cramps or spasms of the stomach, intestines, or bladder, and enuresis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-11285 Supplier Medchemexpress LLC Supplier No. HYB1188S1MG
Propantheline-d3 (bromide) is a deuterium-labeled version of Propantheline bromide. Propantheline bromide is an antimuscarinic agent used to treat hyperhidrosis, cramps or spasms of the stomach, intestines, or bladder, and enuresis.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have potential to affect pharmacokinetic and metabolic profiles of drugs.