Learn More
Medchemexpress LLC Propantheline-d3 bromide | 64717-35-7 | 99.0% | 5 MG
SDP
Catalog No. 5000413626
Encompass_Preferred
Propantheline-d3 bromide is the deuterium labeled Propantheline bromide, functioning as an antimuscarinic agent. It is used for treating conditions like hyperhidrosis, cramps or spasms of the stomach, intestines, or bladder, and enuresis. This compound can also serve as a tracer or an internal standard in quantitative analysis, benefiting from deuterium substitution which can impact pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Functions as an antimuscarinic agent
  • Used in the treatment of hyperhidrosis, cramps, spasms, and enuresis
  • Can be utilized as a tracer
  • Suitable as an internal standard for NMR, GC-MS, or LC-MS quantitative analysis
  • Incorporates stable heavy isotopes for drug development quantitation
Catalog No. 50-004-13626 Supplier Medchemexpress LLC Supplier No. HYB1188S5MG
Propantheline-d3 bromide is the deuterium labeled Propantheline bromide, functioning as an antimuscarinic agent. It is used for treating conditions like hyperhidrosis, cramps or spasms of the stomach, intestines, or bladder, and enuresis. This compound can also serve as a tracer or an internal standard in quantitative analysis, benefiting from deuterium substitution which can impact pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled compound
  • Functions as an antimuscarinic agent
  • Used in the treatment of hyperhidrosis, cramps, spasms, and enuresis
  • Can be utilized as a tracer
  • Suitable as an internal standard for NMR, GC-MS, or LC-MS quantitative analysis
  • Incorporates stable heavy isotopes for drug development quantitation