Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propantheline-d3 bromide | 64717-35-7 | MFCD01311246 | 99.0% | 451.41 g/mol | C23H27D3BrNO3 | 10 MG
SDP

Catalog No. 5000297749
Encompass_Preferred

Deuterium-labeled propantheline bromide provided as a high-purity solid for use as a tracer and internal standard in quantitative mass spectrometry and NMR applications.

  • Deuterium-labeled tracer for MS and NMR quantitative analysis.
  • High purity: 99.0%.
  • Molecular formula: C23H27D3BrNO3; molecular weight: 451.41 g/mol.
  • Appearance: off-white to light yellow solid.
  • Solubility: DMSO ≥ 100 mg/mL; H2O 50 mg/mL (may require ultrasonic).
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small analytical quantities, e.g., 10 mg pack.

Catalog No. 50-002-97749 Supplier Medchemexpress LLC Supplier No. HYB1188S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled propantheline bromide provided as a high-purity solid for use as a tracer and internal standard in quantitative mass spectrometry and NMR applications.

  • Deuterium-labeled tracer for MS and NMR quantitative analysis.
  • High purity: 99.0%.
  • Molecular formula: C23H27D3BrNO3; molecular weight: 451.41 g/mol.
  • Appearance: off-white to light yellow solid.
  • Solubility: DMSO ≥ 100 mg/mL; H2O 50 mg/mL (may require ultrasonic).
  • Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small analytical quantities, e.g., 10 mg pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.