Learn More
Apexbio Technology LLC Proparacaine HCl 5875-06-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062049
Encompass_Preferred
Proparacaine HCl (CAS 5875-06-9) is a small-molecule antagonist of voltage-gated sodium channels By inhibiting sodium ion entry through these channels Proparacaine HCl effectively suppresses action potential generation and neuronal signaling Assays demonstrate an ED50 value of approximately 3 4 mM Due to its sodium channel inhibition Proparacaine HCl is utilized in scientific research to study neuronal excitability ion channel function and related electrophysiological processes
Catalog No. 50-000-62049 Supplier Apexbio Technology LLC Supplier No. B22745.1
Proparacaine HCl (CAS 5875-06-9) is a small-molecule antagonist of voltage-gated sodium channels By inhibiting sodium ion entry through these channels Proparacaine HCl effectively suppresses action potential generation and neuronal signaling Assays demonstrate an ED50 value of approximately 3 4 mM Due to its sodium channel inhibition Proparacaine HCl is utilized in scientific research to study neuronal excitability ion channel function and related electrophysiological processes