Learn More
Selleck Chemical LLC Proparacaine HCl S1828-50mg
SDP
Catalog No. 503109131
Encompass_Preferred
Proparacaine HCl(Proxymetacaine Hydrochloride) is a voltage-gated sodium channels antagonist with ED50 of 3 4 mM
Catalog No. 50-310-9131 Supplier Selleck Chemical LLC Supplier No. S182850MG
Add to Cart
Add to Cart
Proparacaine HCl(Proxymetacaine Hydrochloride) is a voltage-gated sodium channels antagonist with ED50 of 3 4 mM