Learn More
Medchemexpress LLC Proparacaine hydrochloride | 5875-06-9 | 99.9% | C16H27ClN2O3 | 1 ML
SDP
Catalog No. 5000433827
Encompass_Preferred
Proparacaine hydrochloride, also known as Proxymetacaine hydrochloride, is a derivative of lidocaine. It exhibits immunomodulatory and glucocorticomimetic activity. This product is intended for research use only.
  • Exhibits dose- and time-dependent cytotoxicity to HCS cells
  • Induces apoptosis of HCS cells
  • Induces G1 phase arrest
  • Activates caspase-2, -3, and -9
  • Modulates Bcl-xL and Bax expression
  • Reduces nasal symptoms and inflammation in allergic rhinitis mouse models
Catalog No. 50-004-33827 Supplier Medchemexpress LLC Supplier No. HY6601210MM/1ML
Proparacaine hydrochloride, also known as Proxymetacaine hydrochloride, is a derivative of lidocaine. It exhibits immunomodulatory and glucocorticomimetic activity. This product is intended for research use only.
  • Exhibits dose- and time-dependent cytotoxicity to HCS cells
  • Induces apoptosis of HCS cells
  • Induces G1 phase arrest
  • Activates caspase-2, -3, and -9
  • Modulates Bcl-xL and Bax expression
  • Reduces nasal symptoms and inflammation in allergic rhinitis mouse models