Learn More
Medchemexpress LLC Proparacaine hydrochloride | 5875-06-9 | 99.9% | C16H27ClN2O3 | 500 MG
SDP
Catalog No. 5000433817
Encompass_Preferred
Proparacaine hydrochloride (Proxymetacaine hydrochloride) is a derivative of lidocaine, with immunomodulatory effect and glucocorticomimetic activity. It is for research use only.
  • Immunomodulatory effects
  • Glucocorticomimetic activity
  • Induces apoptosis of HCS cells
  • Decreases nasal symptoms, eosinophils, goblet cells, and mast cells in allergic rhinitis mouse models
  • Reduces nasal rubbing and sneezing in allergic rhinitis mouse models
Catalog No. 50-004-33817 Supplier Medchemexpress LLC Supplier No. HY66012500MG
Proparacaine hydrochloride (Proxymetacaine hydrochloride) is a derivative of lidocaine, with immunomodulatory effect and glucocorticomimetic activity. It is for research use only.
  • Immunomodulatory effects
  • Glucocorticomimetic activity
  • Induces apoptosis of HCS cells
  • Decreases nasal symptoms, eosinophils, goblet cells, and mast cells in allergic rhinitis mouse models
  • Reduces nasal rubbing and sneezing in allergic rhinitis mouse models