Learn More
Medchemexpress LLC Proparacaine hydrochloride | 5875-06-9 | C16H27ClN2O3 | 1 G
SDP
Catalog No. 5000434247
Encompass_Preferred
Proparacaine hydrochloride, also known as Proxymetacaine Hydrochloride, is a derivative of lidocaine. It exhibits immunomodulatory and glucocorticomimetic activities.
  • Exhibits immunomodulatory and glucocorticomimetic activities
  • Demonstrates dose- and time-dependent cytotoxicity to HCS cells in vitro
  • Induces apoptosis and G1 phase arrest in HCS cells
  • Activates caspase-2, -3, and -9 pathways
  • Disrupts mitochondrial transmembrane potential
  • Modulates Bcl-xL and Bax expression
  • Increases cytoplasmic cytochrome C and apoptosis-inducing factor
  • Reduces nasal symptoms in allergic rhinitis mouse models
Catalog No. 50-004-34247 Supplier Medchemexpress LLC Supplier No. HY660121G
Proparacaine hydrochloride, also known as Proxymetacaine Hydrochloride, is a derivative of lidocaine. It exhibits immunomodulatory and glucocorticomimetic activities.
  • Exhibits immunomodulatory and glucocorticomimetic activities
  • Demonstrates dose- and time-dependent cytotoxicity to HCS cells in vitro
  • Induces apoptosis and G1 phase arrest in HCS cells
  • Activates caspase-2, -3, and -9 pathways
  • Disrupts mitochondrial transmembrane potential
  • Modulates Bcl-xL and Bax expression
  • Increases cytoplasmic cytochrome C and apoptosis-inducing factor
  • Reduces nasal symptoms in allergic rhinitis mouse models