Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Proparacaine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P21565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21565G 5 g
P21561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21565G Supplier TCI America Supplier No. P21565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6-9-5875
Molecular Formula C16H27ClN2O3
Molecular Weight (g/mol) 330.85
MDL Number MFCD00083467
InChI Key BFUUJUGQJUTPAF-UHFFFAOYSA-N
Synonym Proxymetacaine Hydrochloride, 2-(Diethylamino)ethyl 3-Amino-4-propoxybenzoate Hydrochloride
PubChem CID 517321
IUPAC Name 2-(diethylamino)ethyl 3-amino-4-propoxybenzoate;hydrochloride
SMILES CCCOC1=C(C=C(C=C1)C(=O)OCCN(CC)CC)N.Cl

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 5 g
Formula Weight 330.85
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Proparacaine Hydrochloride
RTECSNumber DG3065000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.