Learn More
TARGETMOL CHEMICALS INC PROPARGITE 5MG
SDP
Catalog No. 502580030
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Propargite is a pesticide used to kill mites. Propargite induces (beta)-cell necrosis preceded by DNA damage. Propargite induces MIN6 cell death with an IC 50 of 1 uM [1] . purity: 98%
Catalog No. 50-258-0030 Supplier TARGETMOL CHEMICALS INC Supplier No. T398495MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Propargite is a pesticide used to kill mites. Propargite induces (beta)-cell necrosis preceded by DNA damage. Propargite induces MIN6 cell death with an IC 50 of 1 uM [1] . purity: 98%