Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propargyl acrylate

Catalog No. 54313625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-313-625G 25 g
54-313-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-313-625G Supplier Sigma Aldrich Supplier No. 54313625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Propargyl acrylate is a propargyl ester of acrylic acid. it participates in the preparation of lipid nanoparticles for use in sirna delivery.propargyl acrylate may be used to synthesize:
  • Linear poly(n-isopropylacrylamide)(pnipam)
  • 1-[3-(2-methyl-2-dodecylsulfanylthiocarbonylsulfanylpropionyloxy)propyl]-1h-[1,2,3]triazol-4-ylmethyl acrylate, an acryloyl trithiocarbonate chain transfer agent
  • Propranolol-imprinted core-shell nanoparticles (mipcs)

Specifications

CAS 10477-47-1
Density 0.997 g/mL (at 25°C (literature))
Boiling Point 142°C to 143°C (lit.)
Molecular Formula C6H6O2
Refractive Index n20/D 1.447 (literature)
Linear Formula CH2=CHCO2CH2C///CH
MDL Number MFCD00078451
Quantity 25 g
Synonym Acrylic acid propargyl ester
Molecular Weight (g/mol) 110.11
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.