Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propargyl bromide solution

Catalog No. 8183150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-831-50ML 50 mL
81-831-250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81-831-50ML Supplier Sigma Aldrich Supplier No. 8183150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The product is 80% solution of propargyl bromide in toluene. propargyl bromide (3-bromopropyne, c3h3br, or 3bp) possess propargyl group, which is an important three-carbon building block. it participates in the stereoselective propargylation reaction of α-alkoxy aldehydes. it is used to compose a soil fumigant trizone.propargyl bromide solution may be used in the synthesis of the following:
  • New arabinogalactan propargyl ethers with degree of substitution (ds) up to 2.8, by propargylation of arabino-3,6-galactan
  • Conjugated polyelectrolyte with polyacetylene as the backbone and pyridinium as side groups, poly(propargyl pyridinium bromide)
  • Benzodiazepine derivative, 4-phenyl-1-(prop-2-yn-1-yl)-1h-1,5-benzodiazepin-2(3h)-one
  • N(3)-propargylated 2α-deoxyuridine, which can be encorporated in the DNA
  • Chiral oxygenated acyclic natural products, via distereoselective propargylation of α-hydroxy aldehydes

Specifications

CAS 106-96-7
Quantity 50 mL
Molecular Formula C3H3Br
Linear Formula HC///CCH2Br
MDL Number MFCD00000241
Synonym 3-Bromo-1-propyne
Molecular Weight (g/mol) 118.96
Density 1.38 g/mL (at 20°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.