Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propargyl bromide solution

Catalog No. P51001625G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
125 g
625 g
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P51001625G 625 g
P510015KG 5 kg
P51001125G 125 g
P5100150G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. P51001625G Supplier Sigma Aldrich Supplier No. P51001625G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to make propargylic amines employed in enyne metathesis.Employed in the propargylation of spiro ketones, allylic alcohols, and enone complexes.

Specifications

CAS 106-96-7
Density 1.335 g/mL (at 25°C)
Boiling Point 88°C to 90°C
Molecular Formula C3H3Br
Refractive Index n20/D 1.494
Linear Formula HC///CCH2Br
MDL Number MFCD00000241
Quantity 625 g
Synonym 3-Bromo-1-propyne
Molecular Weight (g/mol) 118.96
Biological Activity Central Nervous System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.