Learn More
Medchemexpress LLC HY-126511 100mg Medchemexpress, Propargyl-C1-NHS ester CAS:132178-37-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY126511100MG
Encompass_Preferred
Medchemexpress, HY-126511 100mg Propargyl-C1-NHS ester CAS:132178-37-1 Propargyl-C1-NHS ester is a nonclaevable linker for antibody-drug-conjugation (ADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1113