Learn More
Medchemexpress LLC Propargyl-C1-NHS ester | 132178-37-1 | MFCD22577791 | 95.1% | 195.17 g/mol | C9H9NO4 | 50 MG
SDP
Catalog No. 5000202958
Encompass_Preferred
Propargyl-C1-NHS ester is an alkyne-containing N-hydroxysuccinimide (NHS) ester used for bioconjugation and click chemistry. It reacts with primary amines to form stable amide bonds and is commonly applied as a non-cleavable linker in antibody-drug conjugate synthesis, labeling, and other conjugation workflows. The material is supplied as a solid and should be stored at -20 °C, sealed and protected from moisture and light.
  • Alkyne-functionalized NHS ester for click chemistry conjugation.
  • Reacts with primary amines to form stable amide bonds.
  • Non-cleavable linker appropriate for antibody-drug conjugates.
  • High purity suitable for research applications.
  • Store at -20 °C, sealed, and protected from moisture and light.
Catalog No. 50-002-02958 Supplier Medchemexpress LLC Supplier No. HY12651150MG
Propargyl-C1-NHS ester is an alkyne-containing N-hydroxysuccinimide (NHS) ester used for bioconjugation and click chemistry. It reacts with primary amines to form stable amide bonds and is commonly applied as a non-cleavable linker in antibody-drug conjugate synthesis, labeling, and other conjugation workflows. The material is supplied as a solid and should be stored at -20 °C, sealed and protected from moisture and light.
  • Alkyne-functionalized NHS ester for click chemistry conjugation.
  • Reacts with primary amines to form stable amide bonds.
  • Non-cleavable linker appropriate for antibody-drug conjugates.
  • High purity suitable for research applications.
  • Store at -20 °C, sealed, and protected from moisture and light.