Learn More
TARGETMOL CHEMICALS INC Propargyl-C1-NHS ester 50MG
SDP
Catalog No. 502878448
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Propargyl-C1-NHS ester (4-Pentynoic Acid Succinimidyl Ester) is a nonclaevable linker used for antibody-drug-conjugation (ADC). Purity 99.76%
Catalog No. 50-287-8448 Supplier TARGETMOL CHEMICALS INC Supplier No. T1856050MG
Also available in 5 mg, 10 mg, 25 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Propargyl-C1-NHS ester (4-Pentynoic Acid Succinimidyl Ester) is a nonclaevable linker used for antibody-drug-conjugation (ADC). Purity 99.76%