Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-C2-NHS ester | 906564-59-8 | 97.0% | 209.20 | C10H11NO4 | 250 MG
SDP

Catalog No. 5000203746
Encompass_Preferred

Propargyl-C2-NHS ester is a noncleavable linker and click-chemistry reagent containing a terminal alkyne (propargyl) group used to introduce an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC) bioconjugation and in antibody-drug conjugate (ADC) linker synthesis.

  • Purity 97.0%.
  • Molecular weight 209.20 g/mol.
  • Chemical formula C10H11NO4.
  • Appearance colorless to off-white solid-liquid mixture.
  • Storage pure form: -20°C (3 years); in solvent or stock solution: -80°C (6 months) or -20°C (1 month).
  • Available sizes 50 mg to 50 g, including a 250 mg quantity.

Catalog No. 50-002-03746 Supplier Medchemexpress LLC Supplier No. HY126512250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-C2-NHS ester is a noncleavable linker and click-chemistry reagent containing a terminal alkyne (propargyl) group used to introduce an alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC) bioconjugation and in antibody-drug conjugate (ADC) linker synthesis.

  • Purity 97.0%.
  • Molecular weight 209.20 g/mol.
  • Chemical formula C10H11NO4.
  • Appearance colorless to off-white solid-liquid mixture.
  • Storage pure form: -20°C (3 years); in solvent or stock solution: -80°C (6 months) or -20°C (1 month).
  • Available sizes 50 mg to 50 g, including a 250 mg quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.