Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-C2-NHS ester | 906564-59-8 | 97.0% | 209.20 g/mol | C10H11NO4 | 500 MG
SDP

Catalog No. 5000202962
Encompass_Preferred

Propargyl-C2-NHS ester is a non-cleavable linker and click-chemistry reagent used for antibody-drug conjugation and general bioconjugation. It features an alkyne (propargyl) moiety for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and an N-hydroxysuccinimide (NHS) ester for efficient amine coupling.

  • Molecular formula: C10H11NO4
  • Molecular weight: 209.20 g/mol
  • Purity: 97.0%
  • Physical form: Colorless to off-white, solid/liquid mixture
  • Solubility: 100 mg/mL in DMSO
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months)
  • Applications: antibody-drug conjugation and general bioconjugation

Catalog No. 50-002-02962 Supplier Medchemexpress LLC Supplier No. HY126512500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-C2-NHS ester is a non-cleavable linker and click-chemistry reagent used for antibody-drug conjugation and general bioconjugation. It features an alkyne (propargyl) moiety for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and an N-hydroxysuccinimide (NHS) ester for efficient amine coupling.

  • Molecular formula: C10H11NO4
  • Molecular weight: 209.20 g/mol
  • Purity: 97.0%
  • Physical form: Colorless to off-white, solid/liquid mixture
  • Solubility: 100 mg/mL in DMSO
  • Storage: pure form -20°C (3 years); in solvent -80°C (6 months)
  • Applications: antibody-drug conjugation and general bioconjugation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.