Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl Chloride (70% in Toluene, ca. 9.2mol/L), TCI America™
SDP

Catalog No. P127325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P127325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P127325G Supplier TCI America Supplier No. P127325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 624-65-7
Molecular Formula C3H3Cl
Molecular Weight (g/mol) 74.51
MDL Number MFCD00000980
InChI Key LJZPPWWHKPGCHS-UHFFFAOYSA-N
Synonym propargyl chloride, 3-chloropropyne, 3-chloro-1-propyne, 1-propyne, 3-chloro, 2-propynyl chloride, propyne, 3-chloro, propargylchloride, unii-3m62yfl252, gamma-chloroallylene, 1-chloro-2-propyne
PubChem CID 12221
IUPAC Name 3-chloroprop-1-yne
SMILES ClCC#C

Specifications

Boiling Point 63°C
Quantity 25 g
UN Number 3286
Formula Weight 74.51
Physical Form Liquid
Chemical Name or Material Propargyl Chloride (70% in Toluene, ca. 9.2mol/L)
EINECSNumber (2)-3300
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.