Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propargyl chloride

Catalog No. 1439955G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-399-55G 5 g
14-399-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-399-55G Supplier Sigma Aldrich Supplier No. 1439955G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Propargyl chloride is an efficient propargylating agent
  • It generates propargyl radicals by the pulsed laser photolysis.propargyl chloride was used to study emission spectra of dissociating propargyl chloride upon photoexcitation at 199 nm
  • It was used in the synthesis of novel tetraaryl analog of bisphenol a and triaryl analog of bisphenol a propargyl resins

Specifications

CAS 624-65-7
Density 1.03 g/mL (at 25°C (literature))
Boiling Point 58°C (lit.)
Molecular Formula C3H3Cl
Refractive Index n20/D 1.435 (literature)
Linear Formula HC///CCH2Cl
MDL Number MFCD00000980
Quantity 5 g
Synonym 3-Chloro-1-propyne
Molecular Weight (g/mol) 74.51
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.