Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Propargyl chloroformate

Catalog No. 4609235ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-092-35ML 5 mL
46-092-325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-092-35ML Supplier Sigma Aldrich Supplier No. 4609235ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Propargyl chloroformate is a propargyl ester. it serves as a protecting group for the hydroxy and amino functional group. rates of solvolysis of propargyl chloroformate has been analyzed in various solvents of widely varying nucleophilicity and ionizing power values using the extended grunwald-winstein equation.propargyl chloroformate may be used for the preparation of the following:
  • Binaphthol-diacetylene derivatives
  • Propargyl pentafluorophenyl carbonate
  • N-poc protected amino acids (poc = propargyloxycarbonyl)

Specifications

CAS 35718-08-2
Density 1.215 g/mL (at 25°C (literature))
Boiling Point 118°C to 122°C (lit.)
Molecular Formula C4H3ClO2
Refractive Index n20/D 1.435 (literature)
Linear Formula HC///CCH2OCOCl
MDL Number MFCD00270127
Quantity 5 mL
Molecular Weight (g/mol) 118.52
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.