Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-O-C1-amido-PEG4-C2-NHS ester | 2101206-92-0 | 95.0% | 458.46 | C20H30N2O10 | 25 MG
SDP

Catalog No. 5000201893
Encompass_Preferred

Propargyl-O-C1-amido-PEG4-C2-NHS ester is a non-cleavable PEG4-based NHS ester linker with a terminal propargyl (alkyne) group used for copper-catalyzed azide-alkyne cycloaddition (CuAAC) bioconjugation and antibody-drug conjugate synthesis.

  • Non-cleavable PEG4 NHS ester linker.
  • Terminal alkyne (propargyl) for CuAAC click chemistry.
  • Suitable for antibody-drug conjugate synthesis and general bioconjugation.
  • Molecular weight 458.46; formula C20H30N2O10; CAS 2101206-92-0.
  • Purity 95.0%.

Catalog No. 50-002-01893 Supplier Medchemexpress LLC Supplier No. HY12651525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-O-C1-amido-PEG4-C2-NHS ester is a non-cleavable PEG4-based NHS ester linker with a terminal propargyl (alkyne) group used for copper-catalyzed azide-alkyne cycloaddition (CuAAC) bioconjugation and antibody-drug conjugate synthesis.

  • Non-cleavable PEG4 NHS ester linker.
  • Terminal alkyne (propargyl) for CuAAC click chemistry.
  • Suitable for antibody-drug conjugate synthesis and general bioconjugation.
  • Molecular weight 458.46; formula C20H30N2O10; CAS 2101206-92-0.
  • Purity 95.0%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.