Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-O-C1-amido-PEG4-C2-NHS ester | 2101206-92-0 | 95.0% | 458.46 g/mol | C20H30N2O10 | 10 MG
SDP

Catalog No. 5000202457
Encompass_Preferred

Propargyl-O-C1-amido-PEG4-C2-NHS ester is a propargyl-functionalized PEG4 spacer bearing an N-hydroxysuccinimide (NHS) ester. It is used as a bioconjugation reagent for click-chemistry and linker construction, and is reactive toward primary amines. Store cold and dry to preserve NHS ester reactivity.

  • Propargyl (alkyne) functional group for click chemistry.
  • NHS ester reactive toward primary amines for conjugation.
  • Four-unit PEG spacer for increased solubility and spacing.
  • Molecular weight 458.46 g/mol.
  • Purity 95.0%.
  • Recommended storage: pure form at -20°C; in solvent at -80°C or -20°C.

Catalog No. 50-002-02457 Supplier Medchemexpress LLC Supplier No. HY12651510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-O-C1-amido-PEG4-C2-NHS ester is a propargyl-functionalized PEG4 spacer bearing an N-hydroxysuccinimide (NHS) ester. It is used as a bioconjugation reagent for click-chemistry and linker construction, and is reactive toward primary amines. Store cold and dry to preserve NHS ester reactivity.

  • Propargyl (alkyne) functional group for click chemistry.
  • NHS ester reactive toward primary amines for conjugation.
  • Four-unit PEG spacer for increased solubility and spacing.
  • Molecular weight 458.46 g/mol.
  • Purity 95.0%.
  • Recommended storage: pure form at -20°C; in solvent at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.