Learn More
Medchemexpress LLC Propargyl-peg1-acid | 55683-37-9 | 97.0% | 250 MG
SDP
Catalog No. 5000341759
Encompass_Preferred
Propargyl-PEG1-acid is a PEG-based PROTAC linker. It is utilized in the synthesis of BTK-CRBN PROTACs and functions as a click chemistry reagent. This product incorporates an Alkyne group which can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Utilized in the synthesis of BTK-CRBN PROTACs
  • Functions as a click chemistry reagent
  • Incorporates an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-41759 Supplier Medchemexpress LLC Supplier No. 250MGHY130504
Propargyl-PEG1-acid is a PEG-based PROTAC linker. It is utilized in the synthesis of BTK-CRBN PROTACs and functions as a click chemistry reagent. This product incorporates an Alkyne group which can engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Utilized in the synthesis of BTK-CRBN PROTACs
  • Functions as a click chemistry reagent
  • Incorporates an Alkyne group
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc)