Learn More
Medchemexpress LLC Propargyl-PEG1-acid | 55683-37-9 | 97.0% | 5 G
SDP
Catalog No. 5000340339
Encompass_Preferred
Propargyl-PEG1-acid is a PEG-based PROTAC linker, utilized in the synthesis of BTK-CRBN PROTACs, including Ibrutinib-based PROTAC 4 and PROTAC 5. PROTAC 5 is known to degrade BTK and induce degradation of CSK, LYN, and LAT2. This compound also functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules. BTK-CRBN PROTACs are stoichiometric degraders, facilitating BTK protein degradation through the recruitment of cereblon E3 ligases.
  • Peg-based PROTAC linker
  • Used in synthesis of BTK-CRBN PROTACs
  • Functions as a click chemistry reagent
  • Features an alkyne group for CuAAc reactions
  • Degrades BTK protein
  • Induces degradation of CSK, LYN, and LAT2
Catalog No. 50-003-40339 Supplier Medchemexpress LLC Supplier No. HY1305045G
Propargyl-PEG1-acid is a PEG-based PROTAC linker, utilized in the synthesis of BTK-CRBN PROTACs, including Ibrutinib-based PROTAC 4 and PROTAC 5. PROTAC 5 is known to degrade BTK and induce degradation of CSK, LYN, and LAT2. This compound also functions as a click chemistry reagent, featuring an alkyne group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules. BTK-CRBN PROTACs are stoichiometric degraders, facilitating BTK protein degradation through the recruitment of cereblon E3 ligases.
  • Peg-based PROTAC linker
  • Used in synthesis of BTK-CRBN PROTACs
  • Functions as a click chemistry reagent
  • Features an alkyne group for CuAAc reactions
  • Degrades BTK protein
  • Induces degradation of CSK, LYN, and LAT2