Learn More
Medchemexpress LLC Propargyl-PEG1-Boc | 488150-84-1 | 97.0% | 184.23 | 100 MG
SDP
Catalog No. 5000351374
Encompass_Preferred
Propargyl-PEG1-Boc is an alkyl/ether-based PROTAC linker, a click chemistry reagent, utilized in the synthesis of PROTACs. It contains an Alkyne group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Used as a PROTAC linker for synthesizing PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an Alkyne group for CuAAc reactions.
  • Enables reactions with molecules possessing Azide groups.
  • Supports the PROTAC mechanism of action, which involves the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-51374 Supplier Medchemexpress LLC Supplier No. HY140026100MG
Propargyl-PEG1-Boc is an alkyl/ether-based PROTAC linker, a click chemistry reagent, utilized in the synthesis of PROTACs. It contains an Alkyne group enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Used as a PROTAC linker for synthesizing PROTACs.
  • Functions as a click chemistry reagent.
  • Contains an Alkyne group for CuAAc reactions.
  • Enables reactions with molecules possessing Azide groups.
  • Supports the PROTAC mechanism of action, which involves the intracellular ubiquitin-proteasome system to selectively degrade target proteins.