Learn More
Medchemexpress LLC Propargyl-PEG1-Boc | 488150-84-1 | 97.0% | 184.23 | 250 MG
SDP
Catalog No. 5000351568
Encompass_Preferred
Propargyl-PEG1-Boc is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs are composed of two different ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a specific protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Utilizes the ubiquitin-proteasome system for targeted protein degradation
Catalog No. 50-003-51568 Supplier Medchemexpress LLC Supplier No. 250MGHY140026
Propargyl-PEG1-Boc is an alkyl/ether-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. PROTACs are composed of two different ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a specific protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Utilizes the ubiquitin-proteasome system for targeted protein degradation