Learn More
Medchemexpress LLC Propargyl-PEG1-Boc | 488150-84-1 | 97.0% | 184.23 | 500 MG
SDP
Catalog No. 5000349476
Encompass_Preferred
Propargyl-PEG1-Boc is an alkyl/ether-based PROTAC linker. It is used in the synthesis of PROTACs and functions as a click chemistry reagent due to its Alkyne group. This enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only, not for sale to patients.
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in PROTAC synthesis
  • Alkyl/ether-based PROTAC linker
  • Suitable for cancer targeted therapy research
Catalog No. 50-003-49476 Supplier Medchemexpress LLC Supplier No. HY140026500MG
Propargyl-PEG1-Boc is an alkyl/ether-based PROTAC linker. It is used in the synthesis of PROTACs and functions as a click chemistry reagent due to its Alkyne group. This enables it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only, not for sale to patients.
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Used in PROTAC synthesis
  • Alkyl/ether-based PROTAC linker
  • Suitable for cancer targeted therapy research