Learn More
Medchemexpress LLC Propargyl-PEG1-bromide | 18668-74-1 | MFCD00099299 | 98.0% | 163.01 g/mol | C5H7BrO | 100 MG
SDP
Catalog No. 5000282213
Encompass_Preferred
Propargyl-PEG1-bromide is a short polyethylene glycol (PEG) derivative bearing a terminal propargyl (alkyne) group and a bromide leaving group. It is used as a reactive linker in synthetic and chemical biology workflows where a minimal PEG spacer and an alkyne handle enable conjugation and further functionalization. Typical applications include click chemistry, bioconjugation, and assembly of drug-delivery constructs.
  • Contains a terminal propargyl (alkyne) functional group for click reactions.
  • Contains a bromide leaving group for nucleophilic substitution.
  • One-unit PEG spacer provides improved solubility with low steric bulk.
  • Enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Suitable for linker synthesis, bioconjugation, and drug-delivery research.
  • Small molecular weight and compact linker length for precision conjugation.
Catalog No. 50-002-82213 Supplier Medchemexpress LLC Supplier No. HYW190805100MG
Propargyl-PEG1-bromide is a short polyethylene glycol (PEG) derivative bearing a terminal propargyl (alkyne) group and a bromide leaving group. It is used as a reactive linker in synthetic and chemical biology workflows where a minimal PEG spacer and an alkyne handle enable conjugation and further functionalization. Typical applications include click chemistry, bioconjugation, and assembly of drug-delivery constructs.
  • Contains a terminal propargyl (alkyne) functional group for click reactions.
  • Contains a bromide leaving group for nucleophilic substitution.
  • One-unit PEG spacer provides improved solubility with low steric bulk.
  • Enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Suitable for linker synthesis, bioconjugation, and drug-delivery research.
  • Small molecular weight and compact linker length for precision conjugation.