Learn More
Medchemexpress LLC Propargyl-PEG1-bromide | 18668-74-1 | MFCD00099299 | 99.2% | 163.01 g/mol | C5H7BrO | 5 G
SDP
Catalog No. 5000283160
Encompass_Preferred
Propargyl-PEG1-bromide is a small polyethylene glycol (PEG) derivative bearing a terminal propargyl (alkyne) group and a bromide leaving group. It is designed for use as a reactive linker or alkylating reagent in bioconjugation, click chemistry, and drug-delivery research. Supplied as a liquid, it provides a short PEG spacer with an alkyne handle for efficient coupling reactions.
  • Provides terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Contains a bromide leaving group for nucleophilic substitution reactions.
  • Short PEG spacer improves aqueous solubility and reduces steric hindrance.
  • Suitable for conjugation and linker synthesis in medicinal chemistry workflows.
  • Compatible with standard storage recommendations when handled under inert atmosphere.
Catalog No. 50-002-83160 Supplier Medchemexpress LLC Supplier No. HYW1908055G
Propargyl-PEG1-bromide is a small polyethylene glycol (PEG) derivative bearing a terminal propargyl (alkyne) group and a bromide leaving group. It is designed for use as a reactive linker or alkylating reagent in bioconjugation, click chemistry, and drug-delivery research. Supplied as a liquid, it provides a short PEG spacer with an alkyne handle for efficient coupling reactions.
  • Provides terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Contains a bromide leaving group for nucleophilic substitution reactions.
  • Short PEG spacer improves aqueous solubility and reduces steric hindrance.
  • Suitable for conjugation and linker synthesis in medicinal chemistry workflows.
  • Compatible with standard storage recommendations when handled under inert atmosphere.