Learn More
Medchemexpress LLC Propargyl-PEG1-NH2 100mg | 122116-12-5 | 99.13 g/mol | C5H9NO | 100 MG
SDP
Catalog No. 5000228948
Encompass_Preferred
Propargyl-PEG1-NH2 is a short polyethylene glycol linker bearing a terminal propargyl (alkyne) group and a primary amine. It is used as a bifunctional coupling handle in medicinal chemistry and bioconjugation workflows, and is compatible with click chemistry for assembling conjugates such as PROTACs.
  • Short PEG1 linker with terminal alkyne and primary amine.
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (click) conjugations.
  • High purity (98.8%) suitable for research and synthetic applications.
  • Available in small research pack sizes for rapid screening and synthesis.
  • Store at 4°C and protect from light; aliquot prepared solutions to avoid repeated freeze-thaw cycles.
Catalog No. 50-002-28948 Supplier Medchemexpress LLC Supplier No. HY116069100MG
Propargyl-PEG1-NH2 is a short polyethylene glycol linker bearing a terminal propargyl (alkyne) group and a primary amine. It is used as a bifunctional coupling handle in medicinal chemistry and bioconjugation workflows, and is compatible with click chemistry for assembling conjugates such as PROTACs.
  • Short PEG1 linker with terminal alkyne and primary amine.
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (click) conjugations.
  • High purity (98.8%) suitable for research and synthetic applications.
  • Available in small research pack sizes for rapid screening and synthesis.
  • Store at 4°C and protect from light; aliquot prepared solutions to avoid repeated freeze-thaw cycles.