Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Propargyl-PEG1-NH2 250mg | 122116-12-5 | 99.13 | C5H9NO | 250 MG
SDP

Supplier:  Medchemexpress LLC HY116069250MG

Encompass_Preferred

Propargyl-PEG1-NH2 is an alkyne-terminated PEG1 amine linker supplied as a research reagent for chemical biology and bioconjugation applications. It is a low-molecular-weight building block with documented purity and handling guidance.

  • Alkyne-terminated PEG1 amine for click chemistry and bioconjugation.
  • Purity 98.8% as provided by the supplier.
  • Molecular formula C5H9NO; molecular weight 99.13.
  • Available in small pack sizes, including 250 MG.
  • Store at 4°C and protect from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Supplied with a data sheet and safety data sheet for handling and storage information.

Catalog No. 50-002-28946


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.